Pyridine, extra dry 99.5%, AcroSeal®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

339421000 339420010 339420025
339421000.EA 41.6 GBP
339421000. 339420010. 339420025.
Pyridine, extra dry 99.5%, AcroSeal®
Pyridine
Formula: C₅H₅N
Boiling Pt: 115…116 °C (760 mmHg)
Melting Pt: –42 °C
Density: 0.978 g/cm³
Flash Pt: 17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011732
CAS Number: 110-86-1
EINECS: 203-809-9
UN: 1282
ADR: 3,II
Merck Index: 13,08060
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f9c5934940218027dae0d80a1f98b6d5179c0a8&vid=9f2ef46a26e488d366e82869b3acb96652b5dab1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.4 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR