Arsenic standard solution, 1000 mg/L in 5% nitric acid (from As), standard for AAS, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

33559.AE 33559.AP
33559.AEEA 31.3 GBP
33559.AE 33559.AP
Arsenic standard solution, 1000 mg/L in 5% nitric acid (from As), standard for AAS, Specpure®
Standards Cationic Standards Arsenic Standards

Molecular Formula: As in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085309


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR