Potassium cyanate ≥97%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

33356.22 33356.36 33356.A3
33356.22EA 24.8 GBP
33356.22 33356.36 33356.A3
Potassium cyanate ≥97%
Potassium cyanate

Purity: 97% CAS No.: 590-28-3 Molecular Formula: KOCN Formula Weight: 81.12 Physical Form: Crystalline Density: 2.056 Appearance: White Odour: Odourless Melting Point: 700-900° dec. Solubility: Soluble in water. Slightly soluble in alcohol Merck Reference: 14,7625 MDL No.: MFCD00011411

Formula: CKNO
Melting Pt: 700…900 °C (dec.)
Density: 2.056
Storage Temperature: Ambient
MDL Number: MFCD00011411
CAS Number: 590-28-3
EINECS: 209-676-3
Merck Index: 14,7625

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR