Oxalic acid dihydrate 99.5-102.5% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

33262.18 33262.30 33262.A1 33262.A7
33262.18EA 25.6 GBP
33262.18 33262.30 33262.A1 33262.A7
Oxalic acid dihydrate 99.5-102.5% ACS
Oxalic acid dihydrate

Purity: 99.5-102.5% CAS No.: 6153-56-6 Molecular Formula: C2H2O4.2H2O Formula Weight: 126.07 (90.04anhy) Physical Form: Crystalline Density: 1.65 Appearance: White Odour: Odourless pH: 1.3 Melting Point: 101-105° Solubility: Very soluble in water. Moderately soluble in ethanol. Sparingly soluble in ether Beilstein Reference: 3679436 Merck Reference: 14,6911 UN No.: 3261 MDL No.: MFCD00149102


As an analytical reagent, in dye and paint industries, in polymers, in metal treatment, in cleaning and textile finishing, and as a reagent for dehydration and condensation

Formula: HO₂CCO₂H·2H₂O
Boiling Pt: 149…160 °C (1013 hPa)
Melting Pt: 101…105 °C
Density: 1.65
Storage Temperature: Ambient
MDL Number: MFCD00149102
CAS Number: 6153-56-6
EINECS: 205-634-3
UN: 3261
ADR: 8,III
REACH: 01-2119534576-33
Merck Index: 14,6911
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcc1447f5f23a589dc85e10dd090c86e33b6541b&vid=2f0e34543b2f625715b002905fc0dd45fcd75561&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR