Ammonium cerium(IV) nitrate ≥98.5% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Ammonium ceric nitrate, Ammonium hexanitratocerate (IV), Ceric Ammonium Nitrate, Cerium (IV) ammonium nitrate, Diammonium hexanitratocerate

33254.22 33254.36 33254.A3
33254.22EA 93 GBP
33254.22 33254.36 33254.A3
Ammonium cerium(IV) nitrate ≥98.5% ACS
Ammonium cerium (IV) nitrate

Purity: 98.5% min CAS No.: 16774-21-3 Molecular Formula: H8CeN8O18 Formula Weight: 548.23 Physical Form: Granular Appearance: Yellow to orange Odour: Pungent Solubility: Soluble in water and alcohol Merck Reference: 14,1992 UN No.: 1477 MDL No.: MFCD00151121


Analytical chemistry, as an oxidant for organic compounds, as a polymerization catalyst for olefins, and as a scavenger in the manufacture of azides

Formula: (NH₄)₂Ce(NO₃)₆
MW: 548.22 g/mol
Melting Pt: 107…108 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
EINECS: 240-827-6
UN: 1477
ADR: 5.1 II
Merck Index: 14,1992

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR