Zinc stearate (12.5 - 14% ZnO)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

33238.A1 33238.A7 33238.30 33238.18
33238.A1EA 40.7 GBP
33238.A1 33238.A7 33238.30 33238.18
Zinc stearate (12.5 - 14% ZnO)
Zinc stearate

CAS No.: 557-05-1 Molecular Formula: Zn[CH3(CH2)16CO2]2 Formula Weight: 632.48 Physical Form: Powder Appearance: White Melting Point: 130° MDL No.: MFCD00013031

Formula: C₃₆H₇₀O₄Zn
Melting Pt: 130 °C
Density: 1.095 g/cm³ (20 °C)
Flash Pt: 276 °C
Storage Temperature: Ambient
MDL Number: MFCD00013031
CAS Number: 557-05-1
EINECS: 209-151-9
Merck Index: 14,10158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8ad7f3e101681798edbd54e57ad5d2ac82b2e5d&vid=435308cdcb35862fe6cc06384da2ef10d84c5700&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR