Ceramic discs, round, AL23, with hole
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
33140.KF 33147.KF
33140.KFEA 45.3 GBP
33140.KF 33147.KF
Ceramic discs, round, AL23, with hole
Trays/Dishes/Pots Laboratory Trays/Dishes
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.