1,4-Dioxane 99,5 GC_ASSAY_METHOD Stabilised, Puriss. p.a. Reag. Ph. Eur., ACS, ISO analytical reagent, Riedel-de Haën™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

Stabilisation: ~25 mg/L 2,6-di-tert-butyl-4-methylphenol

33147-7L 33147-1L 33147-2.5L 33147-210KG
33147-7LEA 1490 GBP
33147-7L 33147-1L 33147-2.5L 33147-210KG
1,4-Dioxane 99,5 GC_ASSAY_METHOD Stabilised, Puriss. p.a. Reag. Ph. Eur., ACS, ISO analytical reagent, Riedel-de Haën™
1,4-Dioxane

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 102551
RTECS Number: JG8225000
UNSPSC Code: 12191602

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 100…102 °C (1.013 hPa)
Melting Pt: 12 °C
Density: 1.031…1.034 g/cm³ (20 °C)
Flash Pt: 22.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3 II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.5 %
Congealing - Freezing Point Min. 11 °C
Boiling range 100 - 102 °C
Density (D 20/4) 1.031 - 1.034
Refractive index (n 20/D) 1.4210 - 1.4240
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as CH₃COOH) Max. 0.001 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Acetaldehyde Max. 0.005 %
Carbonyl Compounds (as HCHO) Max. 0.01 %
Peroxides (as H₂O₂) Max. 0.005 %
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR