VWR® Bottles, Narrow Neck, PET, Square, with Screw Cap
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
331-0301EA 83.3 GBP
331-0301 VWRI331-0301.1
VWR® Bottles, Narrow Neck, PET, Square, with Screw Cap
Bottles Plastic Bottles
PET, clear, with blue screw cap.
  • Graduated
  • Tamper evident cap (fitted)
  • Lightweight and robust
  • Square
Perfect for sample collection, storage and transport.