Liquid samplers, LiquiSampler
Supplier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5330-2100 5330-2200 5330-1100 5330-1200 5330-1060 5330-2060
331-2150EA 700 GBP
331-2150 331-2151 331-2152 331-2153 331-0036 331-0037
Liquid samplers, LiquiSampler
Samplers Fluid Samplers
Made of ultra-pure and chemically inert PTFE/FEP or PP for contamination-free sampling. Fits all vats and barrels with openings of at least 32 mm Ø.
  • Taking apart and cleaning is extremely simple
  • One hand push button operation
  • Easy to clean − Surfaces are free of pores and crevices, preventing accumulation of dirt
  • Colour coded push buttons in red, blue, green, yellow, white and black
  • Round screw threads proven in foodstuffs hygiene
Suitable for point sampling, cross-sectional sampling, bottom sampling from open and closed barrels, vats, tanks, silosMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable