Crucibles, Tammann, aluminum oxide
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
33052.KF 33053.KF 33055.KF 33056.KF 33058.KF
33052.KFEA 31.2 GBP
33052.KF 33053.KF 33055.KF 33056.KF 33058.KF
Crucibles, Tammann, aluminum oxide
Crucibles
These crucibles according to Tammann are manufactured from aluminium oxide. They display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases. Oxide ceramics are also inert to oxidation and are not subject to radiation damage.