Fish spine beads, AL23
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
32905.KX 32906.KX 32907.KX 32908.KX
32905.KXEA 127 GBP
32905.KX 32906.KX 32907.KX 32908.KX
Fish spine beads, AL23
Beads Ceramic Beads
Pure Al₂O₃ Aluminium oxide, AL 23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.