Epidermal growth factor, recombinant, human, Calbiochem®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

324831-200
324831-200EA 209 GBP
324831-200
Epidermal growth factor, recombinant, human, Calbiochem®
Cell Culture Supplements and Additives

Recombinant, human epidermal growth factor is a protein expressed in E. coli which is identical to human EGF except for an additional N-terminal methionine.


  • Purity: ≥95% by SDS-PAGE
  • Store at –20 °C


It is mitogenic for a wide range of ectoderm- and endoderm-derived cells from tissue such as breast, cornea, epidermis, dermis, liver, pancreas, nerve, amnion, and adrenal medulla. Acts as a survival factor in preventing apoptosis.


Packaging: Packed in plastic ampoule.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR