Anti-Dopamine D 3 Receptor Rabbit Polyclonal Antibody

Supplier: Merck Millipore (Oncogene)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

324402-100
324402-100EA 497 GBP
324402-100
Anti-Dopamine D 3 Receptor Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Dopamine D 3 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR