98,0-100,5 Dry_basis USP, ACS, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

M.O.C. Category R

3222-07EA 0 GBP
3222-07 3222-65
98,0-100,5 Dry_basis USP, ACS, J.T.Baker®
di-Potassium hydrogen phosphate
Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340
MDL Number: MFCD00011383
CAS Number: 7758-11-4
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets U.S.P Requirements 98.0 - 100.5%
GMP Manufactured Product ≤3 ppm
Meets ACS Reagent Chemical Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging ≤0.03%
Bulk Pharmaceutical Chemical ≤0.001%
USP - Assay (K₂HPO₄) (dried basis) 98.0 - 100.5 %
USP - Arsenic (As) ≤ 3 ppm
USP - Carbonate (CO₃) Passes Test
USP - Chloride (Cl) ≤ 0.03 %
USP - Fluoride (F) ≤ 0.001 %
USP - Identification A (USP) Passes Test
USP - Identification B (USP) Passes Test
USP - Insoluble Substances ≤ 0.2 %
USP - Iron (Fe) ≤ 0.003 %
USP - Loss on Drying ≤ 1.0 %
USP - Mono- or Tri- Basic Salt Passes Test
USP - pH (1 in 20) 8.5 - 9.6
USP - Sodium (Na) Passes Test
USP - Sulfate (SO₄) ≤ 0.1 %
Assay (K₂HPO₄) (by acidimetry) ≥ 99.0 %
Chloride (Cl) ≤ 0.003 %
Fluoride (F) ≤ 0.001 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤ 5 ppm
Insoluble Matter ≤ 0.01 %
Trace Impurities - Iron (Fe) ≤ 0.001 %
Loss on Drying at 105°C ≤ 1.0 %
Nitrogen Compounds (as N) ≤ 0.001 %
pH of 5% Solution at 25°C 8.5 - 9.6
Sodium (Na) ≤ 0.05 %
Sulfate (SO₄) ≤ 0.005 %
Trace Impurities - Arsenic (As) ≤ 1 ppm
Trace Impurities - Lead (Pb) ≤ 5 ppm
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Preserve in well closed container.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR