Fehling's reagent I (copper(II) sulphate solution) concentrate for qualitative determination of reducing sugars

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Fehling A solution

31965.264EA 53.7 GBP
31965.264
Fehling's reagent I (copper(II) sulphate solution) concentrate for qualitative determination of reducing sugars
Fehling's reagent I (Copper (II) sulphate solution)

Fehling's reagent solution A


5 ml of solution I + 5 ml of solution II corresponds to about 0,050 g of anhydrous glucose

Formula: CuSO₄
MW: 159.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 1760
ADR: 8 III

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR