Potassium hydroxide 0.5 mol/l (0.5 N) aqueous solution, AVS TITRINORM Volumetric

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Potash caustic, Caustic potash

31950.296EA 25.5 GBP
31950.296
Potassium hydroxide 0.5 mol/l (0.5 N) aqueous solution, AVS TITRINORM Volumetric
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Density: 1.023 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

NIST traceable Confirmed
Titer (20°C; real value 0.2 % accuracy) 0.499 - 0.501 mol/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR