Ceramic tubes, both ends open, thin wall
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
31706.DR 31706.DM 31708.DM 31708.DR 31708.DW 31709.DM 31709.DR 31709.DW
31706.DREA 39.7 GBP
31706.DR 31706.DM 31708.DM 31708.DR 31708.DW 31709.DM 31709.DR 31709.DW
Ceramic tubes, both ends open, thin wall
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.