Tubes, both ends open, ceramic
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
31554.DW 31558.DM 31558.DW 31558.EO 31563.EH 31563.EM 31572.DW 31572.EH 31573.DR 31573.DW 31573.EH 31574.DR 31574.EH 31575.DM 31575.DW 31575.EO
31554.DWEA 43.9 GBP
31554.DW 31558.DM 31558.DW 31558.EO 31563.EH 31563.EM 31572.DW 31572.EH 31573.DR 31573.DW 31573.EH 31574.DR 31574.EH 31575.DM 31575.DW 31575.EO
Tubes, both ends open, ceramic
Tubing Transfer Tubing
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.