Potassium thiocyanate 0.1 mol/l (0.1 N) aqueous solution, AVS TITRINORM Volumetric

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Potassium rhodanide

31481.295EA 58.1 GBP
31481.295
Potassium thiocyanate 0.1 mol/l (0.1 N) aqueous solution, AVS TITRINORM Volumetric
Potassium thiocyanate
Formula: KSCN
MW: 97.18 g/mol
Boiling Pt: 500
Melting Pt: 175
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.0998 - 0.1002 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR