Supelco® Organic Reference Standard, Dextran From Leuconostoc Mesenteroides

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

31424-100MG
31424-100MGEA 188 GBP
31424-100MG
Supelco® Organic Reference Standard, Dextran From Leuconostoc Mesenteroides
Standards Organic Standards

Order Now

SPECIFICATIONS

Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 9004-54-0
MDL No. MFCD00130935
Formula (C₆H₁₀O₅)ₙ
Flash point Not applicable
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR