Potassium permanganate 99, Puriss. p.a. Reag. Ph. Eur., ACS analytical reagent, Fluka™

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Permanganic acid potassium salt

31404-100G 31404-1KG 31404-2.5KG 31404-500G 31404-25KG
31404-100GEA 48.3 GBP
31404-100G 31404-1KG 31404-2.5KG 31404-500G 31404-25KG
Potassium permanganate 99, Puriss. p.a. Reag. Ph. Eur., ACS analytical reagent, Fluka™
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: Dec.
Density: 2.700 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Order Now

Specification Test Results

Assay Min. 99.0 %
Water insoluble matter Max. 0.2 %
Loss on drying (105°C, 2 h) Max. 0.2 %
Chloride (Cl) Max. 200 ppm
Chloride, Chlorate (as Cl) Max. 0.005 %
Sulfate (SO₄) Max. 200 ppm
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR