2-Furaldehyde ≥98% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

31305.A1 31305.30
31305.A1EA 94.1 GBP
31305.A1 31305.30
2-Furaldehyde ≥98% ACS
2-Furaldehyde

Purity: 98% min CAS No.: 98-01-1 Molecular Formula: C5H4O2 Formula Weight: 96.09 Flash Point: 60°(140°F) Physical Form: Liquid Density: 1.159 Refractive Index: 1.5261 Sensitivity: Air Sensitive Appearance: Yellow to brown Odour: Almond-like Melting Point: -38° Boiling Point: 162° Beilstein Reference: 105755 Merck Reference: 14,4304 UN No.: 1199 MDL No.: MFCD00003229

Formula: C₅H₄O₂
Boiling Pt: 162 °C
Melting Pt: –38 °C
Density: 1.159
Flash Pt: 60 °C (140 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003229
CAS Number: 98-01-1
EINECS: 202-627-7
UN: 1199
ADR: 6.1,II
Merck Index: 14,4304

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR