VWR®, Fluids and culture media for sterility testing of pharmaceutical products
Supplier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
311121ZAEA 54.7 GBP
311121ZA 301121ZA 301051ZA 451121ZA 301051PF 563200OA 371121ZA 311122OA 561051ZA 561042TA 401122ZA 301041ZA 555010TA 553560ZA 301050ZK 301120ZK 511122ZA 511052ZA 511121ZA 573560ZA 511120ZA 881054ZA 881124ZA 303200OA 303560ZA 841122ZA 881050ZA 841120ZA 610750ZA 305010TA 311120ZA
VWR®, Fluids and culture media for sterility testing of pharmaceutical products
Media for Microbiology Prepared Media for Microbiology
Culture media for sterility tests, diluents for the homogenisation of samples for the microbiological examination and rinsing fluids for membrane filtration methods.
  • The formulations comply with the requirements of the Harmonised method in the United States (USP), European (EP) and Japanese (JP) Pharmacopoeias
  • Penase (neutralising agent that inactivates penicillin) available for thioglycollate medium
  • Wide mouth flask available for some items (see the descriptions below)
  • Injectable plastic screw inner cap with external protection cap for most of the bottles presented below (see the technical data sheets for details)
  • On request is possible to supply two or three wrapped bottles from 90 ml up to 950 ml, as well as flask with wide-mouth
Fluid K is a diluent for the homogenisation of samples for the microbiological examination.
Fluid A and D are rinsing and/or diluting fluids for membrane filtration methods.
THIOGLYCOLLATE Broth and TRYPTIC SOY Broth (TSB) are highly nutrient liquid medium formulated according to Pharmacopeial harmonised method and used for sterility tests. TSB is used also for pre-enrichment control strains involved in the 'growth promotion' of culture media.
Certifications: The composition of these products are in accordance with the requirements of United States (USP), European (EP) and Japanese (JP) Pharmacopoeias.