dsDNA quantitation kits for the Qubit® fluorometer, AccuGreen™, high-sensitivity and broad-range
Supplier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
31066-T 31066 31068 31068-T 31069 31069-T 31070 31070-T
31066-TEA 131 GBP
31066-T 31066. 31068. 31068-T 31069. 31069-T 31070. 31070-T
dsDNA quantitation kits for the Qubit® fluorometer, AccuGreen™, high-sensitivity and broad-range
Assays Cellular Assays
AccuGreen™ kit is specific for dsDNA and can quantify DNA samples in the range of 0,1 to 10 ng/μl.
  • Optimised for use with the handheld fluorometers such as the Qubit® fluorometer
  • Uses the pre-programmed Qubit® dsDNA high sensitivity assay setting
  • Linear range in assay: 0,01 to 100 ng dsDNA
  • Green fluorescence (Ex/Em 502/523 nm)
  • Storage at 4 °C
Delivery information: Kit contains AccuGreen™ dye 200X, AccuGreen™ buffer 1X, AccuGreen™ standard 1 (0 ng/l) and AccuGreen™ standard 2 (10 ng/l).