Fast probe master mixes
Supplier: Biotium
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
31005 31005-1 31005-2 31016 31016-1 31016-2 31005-T 31016-T
31005.EA 291 GBP
31005. 31005-1 31005-2 31016. 31016-1 31016-2 31005-T 31016-T
Fast probe master mixes
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
A qPCR master mix containing Cheetah™ Taq hotstart DNA polymerase, suitable for probe-based real-time PCR applications.
  • 2X probe-based qPCR master mix for fast cycling protocols
  • Suitable for all fluorescent probe types, including TaqMan®, BHQ®, and Molecular Beacon
  • Contains Cheetah™ Taq hotstart DNA polymerase
Fast Probe Master Mix is designed for fluorescent probe-based PCR applications. It is compatible with probes such as a TaqMan®, BHQ®, and Molecular Beacon. However for most PCR applications we recommend the Forget-Me-Not™ Universal Probe Master Mix, for superior activity.
Fast Probe Master Mix is a 2X master mix, and is supplied with a separate vial of ROX reference dye, for use with high ROX, low ROX, or no ROX instruments.