Iodine 0.05 mol/l (0.1 N) aqueous solution, AVS TITRINORM Volumetric

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

30917.295EA 33.2 GBP
30917.295 30917.320
Iodine 0.05 mol/l (0.1 N) aqueous solution, AVS TITRINORM Volumetric
Iodine
Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
Density: 1.032 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3264
ADR: 8 II
Merck Index: 13,05036

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.0499 - 0.0501 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR