Potassium dichromate 0.04 mol/l (0.24 N) aqueous solution, AVS TITRINORM Volumetric, COD_determination_NFT 90-101

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

30892.298EA 56.7 GBP
30892.298
Potassium dichromate 0.04 mol/l (0.24 N) aqueous solution, AVS TITRINORM Volumetric, COD_determination_NFT 90-101
Potassium dichromate
Formula: K₂Cr₂O₇
MW: 294.18 g/mol
Boiling Pt: 500
Melting Pt: 398
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
UN: 3287
ADR: 6.1 III
Merck Index: 12,07790

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.03992 - 0.04008 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR