Strontium nitrate, anhydrous 98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

30534.30 30534.A1 30534.D9
30534.30EA 22.4 GBP
30534.30 30534.A1 30534.D9
Strontium nitrate, anhydrous 98%
Strontium nitrate

Purity: 98% CAS No.: 10042-76-9 Molecular Formula: N2O6Sr Formula Weight: 211.63 Physical Form: Granular Density: 2.990 Appearance: White Odour: Odourless Melting Point: 570° Solubility: Freely soluble in water. Slightly soluble in alcohol and acetone Merck Reference: 14,8845 UN No.: 1507 MDL No.: MFCD00011248


In railroad flares, signal lights, marine signals

Formula: Sr(NO₃)₂
MW: 211.63 g/mol
Boiling Pt: 645
Melting Pt: 570
Density: 2.990
Storage Temperature: Ambient
MDL Number: MFCD00011248
CAS Number: 10042-76-9
EINECS: 233-131-9
UN: 1507
ADR: 5.1 III
Merck Index: 14,8845

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR