Silver sulphate 6,6 g/l in sulphuric acid, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

30492.241EA 96.5 GBP
30492.241 30492.423
Silver sulphate 6,6 g/l in sulphuric acid, AVS TITRINORM volumetric solution, for COD determination according to NF T 90-101 standard
Silver sulphate
Formula: Ag₂SO₄
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
UN: 1830
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chemical oxygen demand (COD) Passes test
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR