Di-tert-butyl phosphonate ≥95%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

30233.03 30233.06 30233.14
30233.03EA 28.4 GBP
30233.03 30233.06 30233.14
Di-tert-butyl phosphonate ≥95%
Di-tert-butyl phosphonate

Purity: 95% CAS No.: 13086-84-5 Molecular Formula: C8H19O3P Formula Weight: 194.21 Physical Form: Liquid Density: 0.995 Refractive Index: 1.4200 Sensitivity: Air and Moisture Sensitive Boiling Point: 66-68°/0.5mm Solubility: Soluble in organic solvents MDL No.: MFCD00014999


As a solvent, as an antioxidant, and as an intermediate

Formula: C₈H₁₉O₃P
Boiling Pt: 66…68 °C (0.5 mmHg)
Density: 0.995
Storage Temperature: Ambient
MDL Number: MFCD00995019
CAS Number: 13086-84-5
EINECS: 235-996-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR