Hydrochloric acid 5 mol/l (5 N), AVS TITRINORM Volumetric

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

30018.298EA 33.4 GBP
30018.298 30018.447 30018.320 30018.360
Hydrochloric acid 5 mol/l (5 N), AVS TITRINORM Volumetric
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 4.99 - 5.01 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR