Pots with lid, stainless steel
Supplier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
A284-1250 A284-2500 A284-400 A284-5000 A284-650
300-0654EA 90 GBP
300-0654 300-0655 300-0656 300-0657 300-0658
Pots with lid, stainless steel
Trays/Dishes/Pots Pots
These stainless steel pots with lids are general purpose containers that are ideal for use with powders and liquids. The lid is a simple drop on design.
  • 304 stainless steel
  • Hygienic easy clean construction
  • Robust design
  • Easy to use
Ideal for use in food, chemical and general industry.