Disposable samplers, DispoLance
Supplier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5393-5560 5393-5561 5393-5570 5393-5571
300-0263EA 183 GBP
300-0263 300-0264 300-0265 300-0266
Disposable samplers, DispoLance
Samplers Disposable Samplers
The DispoLance samplers can be used for taking large and small sample quantities quickly and easily, directly from bags.
  • Cleanroom manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated
Suitable for free-flowing powders and small-grain granulates.
Order Now
SPECIFICATIONS
Environmentally Preferable