Zone samplers
Supplier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5317-1150 5317-1058 5317-1085 5316-0055 5316-0085 5319-1085 5319-1150 5316-1055 5316-1085 5317-2055 5317-2150 5317-2085 5319-1250 5316-1150 5316-3150 5316-0150 5316-3055 5316-2055 5316-3085 5316-2085 5316-2150 5319-2085 5319-2150 5319-2250
312-1113EA 688 GBP
312-1113 312-1111 312-1112 312-1101 312-1102 312-1121 312-1122 312-1131 312-1132 312-1183 312-1184 312-1185 312-0084 312-0078 312-0082 312-0077 312-0080 312-0079 312-0081 300-0049 300-0050 300-0051 300-0052 300-0053
Zone samplers
Samplers Solid Samplers
For sampling bulk goods, cross-sectional samples can be taken from different sample depths. Suitable for use with a range of sample substrates from fine powder to coarse grain. Multi sampler has three or five separate chambers for taking samples at multiple depths in one action. Uno sampler has one chamber for taking a sample at one specified depth. All layer sampler has no chambers, three or five inlets and this allows one sample to be taken at multiple depths. Jumbo sampler has no chambers and three, five or seven inlets; with a 50 mm tube diameter this is ideal for larger grain sizesMore Product Information