Potassium hexachloroplatinate(IV)

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Dipotassium hexachloroplatinate

295962DEA 0 GBP
295962D
Potassium hexachloroplatinate(IV)
Potassium hexachloroplatinate(IV)
Formula: K₂PtCl₆
MW: 485.99 g/mol
Melting Pt: 250
Density: 3.344 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011389
CAS Number: 16921-30-5
EINECS: 240-979-3
UN: 3290
ADR: 6.1 II
Merck Index: 13,07720

Order Now

Specification Test Results

Assay (calculated as Pt) Min. 39.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR