2,4-Dimethoxytoluene, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning


M-Clarity™ quality level = MQ100

292559-1G 292559-5G
292559-1GEA 86.8 GBP
292559-1G 292559-5G
2,4-Dimethoxytoluene, Sigma-Aldrich®
2,4-Dimethoxytoluene
Formula: C₉H₁₂O₂
Boiling Pt: 110...120 °C/30 mmHg (lit.)
Density: 1,036 g/ml (25 °C) (lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00010749
CAS Number: 38064-90-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d135d6cc0e424d4bc897cb6bd923bd70f2ef06e4&vid=d135d6cc0e424d4bc897cb6bd923bd70f2ef06e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Color) Colorless
Appearance (Form) Liquid
Infrared spectrum Conforms to Structure
Purity (GC) ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR