Zinc sulphate monohydrate ≥98%, purified

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

29245.262EA 110 GBP
29245.262
Zinc sulphate monohydrate ≥98%, purified
Zinc sulphate monohydrate
Formula: ZnSO₄·1H₂O
Density: 3.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149893
CAS Number: 7446-19-7
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 13,10213

Order Now

Specification Test Results

Assay Min. 98 %
Pb (Lead) Max. 100 ppm
Fe (Iron) Max. 0.012 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR