Capillary columns, traditional phases, SLB-IL100, Supelco®
Supplier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
28884-U 28886-U 28887-U
28884-UEA 771 GBP
28884-U 28886-U 28887-U
Capillary columns, traditional phases, SLB-IL100, Supelco®
Chromatography Columns
These are ionic liquid GC columns. They offer a great choice for analysis of neutral and polarisable (contain double and/or triple C-C bonds) analytes.
  • Non-bonded 1,9-di(3-vinylimidazolium)nonane bis(trifluoromethylsulfonyl)imide phase
  • Resistant to damage from moisture/oxygen
  • High thermal stability - maximum temperatures up to 230 °C