2,3,5-Triphenyltetrazolium chloride (TTC), TECHNICAL

Supplier: VWR Chemicals
Ratings:
Avg. Ratings: 3.0 (1 Review)

Total Ratings: 1
Avg. Ratings: out of 5

Warning

Synonyms: Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC

28804.121EA 253 GBP
28804.121
2,3,5-Triphenyltetrazolium chloride (TTC), TECHNICAL
2,3,5-Triphenyltetrazolium chloride
Formula: C₁₉H₁₅ClN₄
Boiling Pt: < 360 °C (1013 hPa)
Melting Pt: 235 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011963
CAS Number: 298-96-4
EINECS: 206-071-6
Merck Index: 13,09815

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR