Toluene ≥99.0%, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

28675.294EA 32.4 GBP
28675.294 28675.328 28675.465 28675.556 28675.950
Toluene ≥99.0%, GPR RECTAPUR®
Toluene
Formula: C₆H₅CH₃
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 01-2119471310-51
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.0 %
Appearance Clear colourless liquid
IR Spectrum Passes test
Boiling point 109 - 112 °C
Density (20/4) 0.865 - 0.868
Benzene Max. 0.02 %
Evaporation residue Max. 50 ppm
Water Max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR