Sodium silicate aqueous solution, TECHNICAL

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Silicic acid sodium salt, Sodium silicate

28079.363EA 103 GBP
28079.363 28079.320 28079.460
Sodium silicate aqueous solution, TECHNICAL
Sodium silicate
Formula: Na₂O(SiO₂)x·xH₂O
Density: 1,34 g/cm³
MDL Number: MFCD00003492
CAS Number: 1344-09-8
EINECS: 215-687-4
UN: 3266
ADR: 8 III
Merck Index: 13,08751

Order Now

Specification Test Results

Silica 25.6 - 27.6 %
Sodium oxide 7.5 - 8.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR