Protein enrichment magnetic beads, Streptavidin Mag Sepharose™
Supplier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
28-9857-99 28-9857-38
28-9857-99EA 364 GBP
28-9857-99 28-9857-38
Protein enrichment magnetic beads, Streptavidin Mag Sepharose™
Beads Magnetic Beads
Streptavidin Mag Sepharose magnetic beads are designed for simple and efficient enrichment of target proteins through immunoprecipitation and purification of biotinylated biomolecules.
  • High-capacity small-scale purification of proteins from cell lysates and biological fluids
  • High purity and yield
  • Easy parallel screening of proteins with high repeatability
  • Simple enrichment of proteins from small or large sample volumes (low µl to high ml scale)
Utilizes strong interaction between biotin and streptavidin ligand immobilized on magnetic beads. MagneticMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable