Affinity chromatography columns, HiTrap™ AVB Sepharose™ HP
Supplier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
28-4112-11
28-4112-11EA 906 GBP
28-4112-11
Affinity chromatography columns, HiTrap™ AVB Sepharose™ HP
Chromatography Columns
AVB Sepharose High Performance is an affinity resin designed for the purification of adeno associated virus (AAV), Prepacked HiTrap columns for simple operation with a syringe, pump, or chromatographic system.
  • One step purification of adeno associated viruses
  • Affinity resin with high selectivity and low background binding
  • Large surface area to give enhanced capacity
AVB Sepharose High performance is an affinity resin with affinity for adeno associated viruses. It enables purification of adeno associated viruses in one step with high purity andMore Product Information