Orthophosphoric acid ≥85.0% (by acidimetry), AR® ACS, Macron Fine Chemicals™

Supplier: Avantor Performance Materials
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

2796-05EA 0 GBP
2796-05 2796-27
Orthophosphoric acid ≥85.0% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Orthophosphoric acid

ACS—Reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Formula: H₃PO₄
Melting Pt: 28 °C
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 85.0 %
ACS - Assay (as H₃PO₄) (by acidimetry) ≥85.0 %
ACS - Antimony (Sb) ≤15.000 ppm
ACS - Arsenic (As) ≤1.000 ppm
ACS - Calcium (Ca) ≤0.002 %
ACS - Chloride (Cl) ≤3 ppm
ACS - Color (APHA) ≤10
ACS - Heavy Metals (as Pb) ≤10 ppm
ACS - Insoluble Matter ≤0.001 %
ACS - Iron (Fe) ≤6 ppm
ACS - Magnesium (Mg) ≤0.002 %
ACS - Manganese (Mn) ≤0.500 ppm
ACS - Nitrate (NO₃) ≤5 ppm
ACS - Potassium (K) ≤6.000 ppm
ACS - Reducing Substances (mL) ≤1.0
ACS - Sodium (Na) ≤10.000 ppm
ACS - Sulfate (SO₄) ≤0.001 %
ACS - Volatile Acids (as CH₃COOH) ≤0.001 %
Product Information (not specifications):
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR