Sodium molybdate(VI) dihydrate ≥99.0%, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate

27936.233EA 169 GBP
27936.233 27936.290 27936.460
Sodium molybdate(VI) dihydrate ≥99.0%, GPR RECTAPUR®
Sodium molybdate(VI) dihydrate
Formula: Na₂MoO₄·2H₂O
MW: 241.95 g/mol
Melting Pt: 687
Density: 3.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718

Order Now

Specification Test Results

Assay Min. 99.0 %
Heavy metals (as Pb) Max. 20 ppm
Cl (Chloride) Max. 50 ppm
NH4 (Ammonium) Max. 20 ppm
SO4 (Sulphate) Max. 0.05 %
Fe (Iron) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR