Sodium carbonate decahydrate Ph. Eur.

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

27761.295EA 69.1 GBP
27761.295 27761.460
Sodium carbonate decahydrate Ph. Eur.
Sodium carbonate decahydrate
Formula: Na₂CO₃·10H₂O
Melting Pt: 33 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149178
CAS Number: 6132-02-1
EINECS: 207-838-8

Order Now

Specification Test Results

Assay (Na2CO3) 36.7 - 40.0 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification (CO3) Passes test
Identification (Na) Passes test
Appearance of solution Passes test
Alkali hydroxides and bicarbonates Passes test
Cl (Chloride) ≤ 50 ppm
SO4 (Sulphate) ≤ 100 ppm
Fe (Iron) ≤ 20 ppm
Heavy metals (as Pb) ≤ 20 ppm
Residual solvents Passes test
As (Arsenic) ≤ 2 ppm
Cd (Cadmium) ≤ 1 ppm
Co (Cobalt) ≤ 1 ppm
Hg (Mercury) ≤ 3 ppm
Ni (Nickel) ≤ 1 ppm
Pb (Lead) ≤ 3 ppm
V (Vanadium) ≤ 1 ppm
Conforms to Ph.Eur. Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR