Adapters with screw thread and standard gound joint cone
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
01311419 01311424 01311429 01311814 01311819 01311824 01312519 01312524 01311829 01312514 01312529 01311414 01313229
275-1122EA 19.1 GBP
275-1122 275-1123 201-1641 201-1642 201-1643 201-1644 275-1129 275-1130 201-1645 201-1031 201-1646 201-1640 201-1647
Adapters with screw thread and standard gound joint cone
Adapters Adapters, Conical Ground Joints
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Threaded adapter with DIN thread and standard ground joint cone.
DIN 12257