Triammonium citrate 97-103%, GPR RECTAPUR®

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

271534HEA 115 GBP
271534H
Triammonium citrate 97-103%, GPR RECTAPUR®
tri-Ammonium citrate
Formula: C₆H₁₇N₃O₇
Melting Pt: 185 °C
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036406
CAS Number: 3458-72-8
EINECS: 222-394-5

Order Now

Specification Test Results

Assay 97 - 103 %
Identification Passes test
pH (20°C; 5 %) 6.0 - 7.5
Ignition residue (SO4) Max. 0.05 %
Cl (Chloride) Max. 100 ppm
Pb (Lead) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR