Clamps for conical standard ground joints, KECK™, DURAN®
Supplier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
286337302 286337405 286337508 286337602
271-7130EA 8.6 GBP
271-7130 271-7138 271-7135 271-7136
Clamps for conical standard ground joints, KECK™, DURAN®
Clamps NS Joint Clamps
Nickel with steel spring. KECK™ conical joint clips ensure a secure connection and thus increase laboratory safety.
  • Easy to fit and do not alter their shape
  • Ideal for applications with very high temperatures